Order Entry
Belgium
ContactUsLinkComponent
Anti-Xiap Rabbit Recombinant Antibody [clone: R02-9A3]
Anti-Xiap Rabbit Recombinant Antibody [clone: R02-9A3]
Catalog # ABNORAB01823.100UL
Supplier:  Abnova
Anti-Xiap Rabbit Recombinant Antibody [clone: R02-9A3]
Catalog # ABNORAB01823.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    Xiap
  • Clonality:
    Recombinant
  • Clone:
    R02-9A3
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against mouse Xiap.
  • Pk:
    100 µl

Specifications

About this item