Anti-BAG6 Rabbit Recombinant Antibody [clone: R04-1C1]
Catalog # ABNORAB01296.100UL
Supplier: Abnova
Specifications
- Antigen symbol:BAG6
- Clonality:Recombinant
- Clone:R04-1C1
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human BAG6.
- Pk:100 µl