Orders LinkContactUsLinkComponent
QuickConnect™ Mop Handles
QuickConnect™ Mop Handles
New Product
Supplier:   CONTEC
QuickConnect™ Mop Handles
Supplier:  CONTEC
New Product
Certificates
QuickConnect™ Mop Handles
QuickConnect™ Mop Handles
6 Options Available Below
Product Details & Documents
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, as well as anodised aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • CONTCFH32
    QuickConnect™ carbon fiber telescopic handle
    QuickConnect™ carbon fiber telescopic handle
    46 - 81 cm (18 - 32")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,00 €
    184,00 €
    1 * 1 ST
       184,00 €
  • CONTCFH18
    QuickConnect™ carbon fiber handle
    QuickConnect™ carbon fiber handle
    46 cm (18")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,20 €
    73,20 €
    1 * 1 ST
       73,20 €
    Specifications:
    New Product
    New Product
  • CONTCFH70
    QuickConnect™ carbon fiber telescopic handle
    QuickConnect™ carbon fiber telescopic handle
    102 - 178 cm (40 - 70")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 189,00 €
    189,00 €
    1 * 1 ST
       189,00 €
  • CONTCFH114
    QuickConnect™ carbon fiber telescopic handle
    QuickConnect™ carbon fiber telescopic handle
    160 - 290 cm (63 - 114")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 276,00 €
    276,00 €
    1 * 1 ST
       276,00 €
  • CONTCFH224
    QuickConnect™ carbon fiber telescopic handle
    QuickConnect™ carbon fiber telescopic handle
    168 - 568 cm (66 - 224")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 522,00 €
    522,00 €
    1 * 1 ST
       522,00 €
  • CONTCFH334
    QuickConnect™ carbon fiber telescopic handle
    QuickConnect™ carbon fiber telescopic handle
    175 - 848 cm (69 - 334")
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 733,00 €
    733,00 €
    1 * 1 ST
       733,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.