Cassette
SIUS PD cassette with HyStream 300kD L: medium channel; single-use/reusable 0,01 m² holder-M
Catalog # SPECXP300MP1L
Supplier: Spectrum Laboratories
Jump to:
Product Details & Documents
SIUS PD cassette with HyStream membrane type and molecular weight cut-off (MWCO) of 300kD.
- medium channel
- Single-use
- Reusable
Specifications
- Description:SIUS PD cassette with HyStream 300kD L: medium channel; single-use/reusable 0,01 m² holder-M