ContactUsLinkComponent
3M™ Clear Reusable Polycon Earplug Storage Box
Catalog # MMMA7100317692
Supplier:  3M
3M™ Clear Reusable Polycon Earplug Storage Box
Catalog # MMMA7100317692
Supplier:  3M
Specifications
  • Description:
    Reusable Carrying case
  • Material:
    PP
  • Pk:
    50
Specifications
About this item
Robust reusable storage case to store 3M™ earplugs safely and securely. Case includes belt clip for convenient storage and easy access. Transparent colour allows for quick visual confirmation that the product is in the box.
  • Can be washed with mild detergent
  • Replace the case only when you need and help reduce wastage
Each storage box has a clip feature allowing you to clip the box on your pocket or belt loop, keeping the box safe, secure and ready to hand. Made from durable polypropylene, this reusable box gives you another packaging option to safely store your 3M™ plugs, such as 3M™ 1271 Earplugs or 3M™ Ultrafit™ Earplugs.