Orders LinkContactUsLinkComponent
Wire Baskets for Test Tubes
Wire Baskets for Test Tubes
Wire Baskets for Test Tubes
Certificates
Wire Baskets for Test Tubes
3 Options Available Below
Product Details & Documents
Wire basket of welded mesh w 8 mm, d 1 mm for test tubes and more. Made up of stainless steel W-no. 1.4301, cube shape, electrochemically polished.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 100×100×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 120×120×120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,40 €
    35,40 €
    1 * 1 ST
       35,40 €
  • 140×140×140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.