Orders LinkContactUsLinkComponent
Experimentation kits, affinity chromatography
Experimentation kits, affinity chromatography
Affinity Chromatography
Catalog # BE-417
Supplier:   G-Biosciences
Experimentation kits, affinity chromatography
Catalog # BE-417
Supplier:  G-Biosciences
Experimentation kits, affinity chromatography
Experimentation kits, affinity chromatography
Affinity Chromatography
Price
168,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The Affinity Chromatography kit teaches the basic principle of affinity chromatography utilising highly specific affinity columns. This lab activity involves preparation of a crude protein extract and running affinity exchange chromatography for isolation of a protein.
  • The principle of affinity chromatography
  • Immobilisation of protein on affinity columns
  • Binding and elution of proteins on affinity columns
  • Factors influencing affinity chromatography
  • Hands-on affinity chromatography lab activity
Also required: Spectrophotometer and cuvettes or microplate reader and microplate (optional), low speed centrifuge for 1,5 to 2 ml tubes.
Delivery information: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.
Documents
Specifications
  • Description:
    Affinity Chromatography