Orders LinkContactUsLinkComponent
Experimentation kits, nucleic acid quantification
Experimentation kits, nucleic acid quantification
Nucleic Acid Quantification
Catalog # BE-318
Supplier:   G-Biosciences
Experimentation kits, nucleic acid quantification
Catalog # BE-318
Supplier:  G-Biosciences
Experimentation kits, nucleic acid quantification
Experimentation kits, nucleic acid quantification
Nucleic Acid Quantification
Price
283,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The kit utilises the principle of diffusion of nucleic acids on a nylon membrane to determine their concentration. No spectrophotometers required.
  • No expensive equipment required
  • Rapidly quantitate DNA and RNA samples
  • Enough reagents for over 50 assays
Also required: DNA samples.
Nucleic acid concentration can be measured by measuring the diameter of diffused nucleic acid spots or comparing the colour density of the spots with a set of known standards. Accurate and fast DNA and RNA concentrations can be determined with as little as 1 μl of DNA sample.
Delivery information: Supplied with 50 assay strips in addition to all the buffers and scales required for DNA or RNA quantification.
Documents
Specifications
  • Description:
    Nucleic Acid Quantification