ContactUsLinkComponent
Experimentation kits, whose fingerprints were left behind?
Experimentation kits, whose fingerprints were left behind?
Catalog # EDVOS-91
Supplier:  EDVOTEK
undefined
Experimentation kits, whose fingerprints were left behind?
Catalog # EDVOS-91
Supplier:  EDVOTEK
Specifications
  • Description:
    Whose Fingerprints Were Left Behind
  • Pk:
    1
Specifications
About this item
Evidence left behind at a crime scene can identify a potential culprit. Even in this age of DNA, fingerprints and blood stains are still important in helping to identify a criminal. In this experiment, students solve a crime by dusting for fingerprints and use fluorescent dust to search for and identify trace amounts of blood.
  • For 10 lab groups
  • Complete in 50 minutes
Also required: Alcohol and longwave UV light.
Delivery: Kit includes instructions, brushes, magnifying lens, fingerprint cards, black dusting powder, fluorescent green and grey dye dusting powder, and fingerprint lifters.