ContactUsLinkComponent
Experimentation kits, principles of gel filtration chromatography
Experimentation kits, principles of gel filtration chromatography
Catalog # EDVO108
Supplier:  EDVOTEK
undefined
Experimentation kits, principles of gel filtration chromatography
Catalog # EDVO108
Supplier:  EDVOTEK
Specifications
  • Description:
    Principles of Gel Filtration Chromatography
  • Pk:
    1
Specifications
About this item
This experiment introduces chromatographic separation to the class and shows students how dyes of different colours separate on the basis of their size and shape. Columns may be rinsed and reused.
  • For 10 lab groups
  • Packing column 20 minutes, column separation 40 minutes
Also required: 50 ml or 100 ml beakers, 25 ml beaker or test tube, ring stands with clamps, and distilled water.
Delivery: Kit includes instructions, sample mixture, chromatography columns, dry matrix, elution buffer, transfer pipettes, and microtest tubes.