ContactUsLinkComponent
Do onions, strawberries and bananas have DNA?
Do onions, strawberries and bananas have DNA?
Catalog # EDVOS-75
Supplier:  EDVOTEK
undefined
Do onions, strawberries and bananas have DNA?
Catalog # EDVOS-75
Supplier:  EDVOTEK
Specifications
  • Description:
    Do Onions, Strawberries and Bananas Have DNA?
  • Pk:
    1
Specifications
About this item
Kit allows students to construct DNA models and then extract DNA from onions, strawberries or bananas.
  • For 10 lab groups
  • Complete in 30 minutes
Also required: 95 to 100% isopropyl alcohol, fruit and vegetables.
Also required: 95-100% isopropyl alcohol, fruit and vegetables.
Delivery: Kit includes instructions, DNA extraction buffer, DNA sample in capped test tube, transfer pipettes, pop beads, glass rod, DNA spooling rods, test tubes, and salt.