ContactUsLinkComponent
Kits: why do people look different?, Ready-to-Load™
Kits: why do people look different?, Ready-to-Load™
Catalog # 763-0007
Supplier:  EDVOTEK
undefined
Kits: why do people look different?, Ready-to-Load™
Catalog # 763-0007
Supplier:  EDVOTEK
Specifications
  • Description:
    Why Do People Look Different?
  • Pk:
    1 KIT
Specifications
About this item
This kit is for teaching students how people's physical traits are a reflection of their genes. In this simulation, students use electrophoresis to separate dyes which represent genetic traits.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.