Orders LinkContactUsLinkComponent
Micropipetting basics
Micropipetting basics
Micropipetting basics
Catalog # EDVOS-44
Supplier:   EDVOTEK
Micropipetting basics
Catalog # EDVOS-44
Supplier:  EDVOTEK
Micropipetting basics
Micropipetting basics
Micropipetting basics
Price
64,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Kit for teaching students how to use a micropipette with ease and accuracy using multi-coloured dyes.
  • 10 lab groups
  • Complete in 45 minutes
Also required: 5 to 50 µl adjustable or 10 µl fixed micropipette and tips.
Delivery information: Kit includes instructions, various coloured dye samples and a pipette card.
Specifications
  • Description:
    Micropipetting basics