ContactUsLinkComponent
TAE buffer, 50X
TAE buffer, 50X
Catalog # EDVO607
Supplier:  EDVOTEK
CAS Number:  
TAE buffer, 50X
Catalog # EDVO607
Supplier:  EDVOTEK
CAS Number:  
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    TAE Buffer (50X)
Specifications
About this item
This 50X concentrated solution (Tris-acetate, EDTA, pH 7,8) is sufficient for making 5 L of diluted working buffer for making agarose gels and electrophoresis chamber buffer.