Order Entry
Belgium
Orders LinkContactUsLinkComponent
Gel loading solution, 10X

Gel loading solution, 10X

Gel Loading Solution (10X), 1 mL
Catalog # EDVO606
Supplier:   EDVOTEK
Gel loading solution, 10X
Catalog # EDVO606
Supplier:  EDVOTEK
CAS Number:  
Gel loading solution, 10X

Gel Loading Solution (10X), 1 mL

CAS Number:  
Price
11,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

1 ml concentrate for 200 samples.

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Gel Loading Solution (10X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.