ContactUsLinkComponent
Experimentation kits, PCR-based Alu-Human DNA typing
Experimentation kits, PCR-based Alu-Human DNA typing
Catalog # EDVO333
Supplier:  EDVOTEK
undefined
Experimentation kits, PCR-based Alu-Human DNA typing
Catalog # EDVO333
Supplier:  EDVOTEK
Specifications
  • Description:
    PCR-based Alu-Human DNA Typing
  • Pk:
    1 KIT
Specifications
About this item
Students use primers for a 300 base pair Alu insertion in chromosome 16 (PV92) to determine their own genotype. They can then compare their class results with others around the world over the internet.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Delivery: Kit includes instructions, proteinase K, PCR beads, control DNA and primers, microtubes, chelating agent, agarose, DNA ladder, practice gel loading solution, gel loading dye, electrophoresis buffer, and gel stain.