ContactUsLinkComponent
Experimentation kits, DNA/RNA microarrays
Experimentation kits, DNA/RNA microarrays
Catalog # EDVO235
Supplier:  EDVOTEK
undefined
Experimentation kits, DNA/RNA microarrays
Catalog # EDVO235
Supplier:  EDVOTEK
Specifications
  • Description:
    DNA/RNA Microarrays
  • Pk:
    1 KIT
Specifications
About this item
Microarray technology allows scientists to screen large numbers of samples in one assay. This technology allows high-throughput screening of small samples and allows comparisons to be made between healthy and unhealthy tissues. In this experiment, students apply simulated DNA and RNA samples to a membrane to screen for positive and negative samples.
  • For 10 lab groups
  • Complete in 60 minutes
Also required: 5 µl fixed volume or adjustable micropipette and tips.
Delivery: Kit includes instructions, simulated patient DNA and RNA samples and controls, microarray cards, plastic bags to incubate membrane, microtest tubes and pipettes.