Orders LinkContactUsLinkComponent
Coloured beads
Coloured beads
Coloured beads
Catalog # EDVO1500
Supplier:   EDVOTEK
Coloured beads
Catalog # EDVO1500
Supplier:  EDVOTEK
Coloured beads
Coloured beads
Coloured beads
Price
64,90 €
Log in for availability
1 * 100 ST
Product Details & Documents
A set of coloured beads that can be designated to represent the Watson-Crick DNA bases: A, T, G, C.
Can be used to demonstrate concepts related to the structure and biology of DNA.
Ordering information: Includes 150 beads of each colour with detailed outline of various demonstrations.
Specifications
  • Description:
    Coloured beads