Orders LinkContactUsLinkComponent
Codesoft Lite Software
Codesoft Lite Software
Supplier:   Brady
Codesoft Lite Software
Supplier:  Brady
Certificates
Codesoft Lite Software
4 Options Available Below
Product Details & Documents
Codesoft Lite label design software can be used for asset tracking, barcode labelling, and inventory and inspection labelling applications.
  • Only available as a single user license
  • Limited to 1 thermal printer
Languages availble include Arabic (Egypt), Czech, Danish, Dutch, English, Finnish, French, German, Hebrew, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Spanish, Swedish, and Turkish.
Compatible with BBP11, BBP12, BBP72, PR Plus, IP Series, PAM3000, PAM3600, THT 76mm Core, and TLS-PC LINK printers.
Product Family Options
Product Information
  • Availability
    Price
  • BRDY196181
    Codesoft lite subscription online
    Codesoft lite subscription online
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 441,00 €
    441,00 €
    1 * 1 ST
       441,00 €
  • BRDY196184
    Codesoft lite subscription USB
    Codesoft lite subscription USB
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 441,00 €
    441,00 €
    1 * 1 ST
       441,00 €
  • BRDY362916
    Codesoft lite prepetual online 3 year SMA
    Codesoft lite prepetual online 3 year SMA
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 914,00 €
    914,00 €
    1 * 1 ST
       914,00 €
  • BRDY362918
    Codesoft lite prepetual USB 3 year SMA
    Codesoft lite prepetual USB 3 year SMA
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 210,00 €
    1 210,00 €
    1 * 1 ST
       1 210,00 €