ContactUsLinkComponent
LMDA (Lee's Multi-Differential Agar)
LMDA (Lee's Multi-Differential Agar)
Catalog # DOEH923555244
Supplier:  DOEHLER GROUP
CAS Number:  
undefined
LMDA (Lee's Multi-Differential Agar)
Catalog # DOEH923555244
Supplier:  DOEHLER GROUP
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    1
  • Description:
    LMDA (Lee's Multi-Differential Agar)
  • Filling:
    500 g
Specifications
About this item
Siebel-Instituts agar medium for the selective detection and quantitative determination of microorganisms (bacteria, yeasts) from the brewing environment.
  • Certified by the Siebel-Institute for technology
  • Ready-to-use powder for easy handling
  • Functionality tested with strains originally isolated from the brewing environment
Identification of the target bacteria is facilated by characteristic colour changes of the medium. In addition, acid-forming bacteria show a typical halo formation due to the breakdown of the calcium carbonate contained in the LMDA medium.