ContactUsLinkComponent
SteriWare Graduated Beakers
SteriWare Graduated Beakers
Catalog # SAMP8008P-50S
Supplier:  Sampling Systems
Supplier #: 8008P-50S
undefined
SteriWare Graduated Beakers
Catalog # SAMP8008P-50S
Supplier:  Sampling Systems
Supplier #: 8008P-50S
Log in for availability
Select an Option
50 ml
Select an Option
SteriWare beaker
Select an Option
02
Select an Option
50
About this item
The SteriWare range of greduated beakers are ideal for use in the production of drugs, chemicals and cosmetics.
  • Made from food grade, FDA and EC compliant PP
  • Individually bagged
  • Choice of sterile and non sterile
  • Wide range of sizes
  • 100% manufactured and packed in a cleanroom
The range of individually bagged graduated beakers from sampling systems are crucial in the production of drugs, chemicals and cosmetics. As they are indivicually bagged they prevent cross contamination and they have graduation marks and a pouring spout to ensure the convnenient handling of liquids.
Frequently Bought Together