Orders LinkContactUsLinkComponent
Double Ended Spoons
Double Ended Spoons
Supplier:   Sampling Systems
Double Ended Spoons
Supplier:  Sampling Systems
Certificates
Double Ended Spoons
Double Ended Spoons
5 Options Available Below
Product Details & Documents
Double ended spoon for taking different sample sizes.
  • Made from stainless steel
  • Ideal for sampling powders and liquids
  • Various sizes available
These double ended spoons are great for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary allowing for flexibility in sampling both powders and liquids.
Product Family Options
Product Information
  • Length
    Capacity
    Material
    Availability
    Price
  • SAMPA104-210
    Double ended spoon
    Double ended spoon
    210 mm
    1,6/3 ml
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,00 €
    19,00 €
    1 * 1 ST
       19,00 €
  • SAMPA104-250
    Double ended spoon
    Double ended spoon
    250 mm
    1,8/5 ml
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,60 €
    22,60 €
    1 * 1 ST
       22,60 €
  • SAMPA104-300
    Double ended spoon
    Double ended spoon
    300 mm
    2/8 ml
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,10 €
    27,10 €
    1 * 1 ST
       27,10 €
  • SAMPA104-150
    Double ended spoon
    Double ended spoon
    150 mm
    0,5/2 ml
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,60 €
    13,60 €
    1 * 1 ST
       13,60 €
  • SAMPA104-180
    Double ended spoon
    Double ended spoon
    180 mm
    0,8/2,8 ml
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,30 €
    16,30 €
    1 * 1 ST
       16,30 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.