Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tris-Glycine buffer

Tris-Glycine buffer

Supplier:   EDVOTEK
Tris-Glycine buffer
Supplier:  EDVOTEK
Certificates
Tris-Glycine buffer

2 Options Available Below

Product Details & Documents

For vertical polyacrylamide gel electrophoresis.

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • EDVO655
    Tris-glycine-SDS buffer (10X for 5 l)
    Tris-glycine-SDS buffer (10X for 5 l)
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 27,50 €
    27,50 €
    1 * 500 mL
       27,50 €

  • EDVO656
    Tris-glycine buffer (10X for 5 l)
    Tris-glycine buffer (10X for 5 l)
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 27,50 €
    27,50 €
    1 * 500 mL
       27,50 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.