Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Supplier:   EDVOTEK
Pipette tips
Supplier:  EDVOTEK
Certificates
Pipette tips
7 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Tip style
    Volume
    Tip colour
    Packed
    Availability
    Price
  • Ultra
    0,5 - 10 µl
    Transparent
    2 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 192 ST
    1 * 192 ST - 16,50 €
    16,50 €
    1 * 192 ST
       16,50 €
  • Standard
    200 - 1000 µl
    Blue
    2 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 192 ST
    1 * 192 ST - 20,90 €
    20,90 €
    1 * 192 ST
       20,90 €
  • Standard
    1 - 200 µl
    Yellow
    Bag of 1000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 49,50 €
    49,50 €
    1 * 1.000 ST
       49,50 €
  • Standard
    200 - 1000 µl
    Blue
    Bag of 1000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 66,00 €
    66,00 €
    1 * 1.000 ST
       66,00 €
  • Standard
    1 - 200 µl
    Yellow
    2 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 192 ST
    1 * 192 ST - 16,50 €
    16,50 €
    1 * 192 ST
       16,50 €
  • Fine tip
    1 - 200 µl
    Transparent
    2 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 204 ST
    1 * 204 ST - 38,50 €
    38,50 €
    1 * 204 ST
       38,50 €
  • Ultra
    0,5 - 10 µl
    Transparent
    Bag of 1000
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 66,00 €
    66,00 €
    1 * 1.000 ST
       66,00 €