Single channel pipettes, variable volume
Supplier: EDVOTEK
Aller à :
À propos de cet article
Sturdily designed handheld pipettes with volumes ranging from 0,1 to 5000 µl. They are easy to use, accurate and use standard micropipette tips.
- Volume easily selected by twisting the top
- Lightweight design and tip ejector makes operation fast and easy
- Tool and instructions included for self-calibration
8 Options disponibles ci-dessous
Product Details & Documents
Sturdily designed handheld pipettes with volumes ranging from 0,1 to 5000 µl. They are easy to use, accurate and use standard micropipette tips.
- Volume easily selected by twisting the top
- Lightweight design and tip ejector makes operation fast and easy
- Tool and instructions included for self-calibration
Product Family Options
Informations sur le produit
- VolumeAvailabilityPrice