ContactUsLinkComponent
96 Well Plate, RCH PWCX
96 Well Plate, RCH PWCX
  TCAT675-0201RCH
 :  Tecan
96 Well Plate, RCH PWCX
  TCAT675-0201RCH
 :  Tecan
  • Description:
    Plate RCH PWCX, 1 ml, 19 µm
  • Phase:
    PWCX
  • Filling:
    20 mg
  • Pk:
    1
Polymeric, weak cation exchange chemistry for strong bases.
  • Ion-exchange
  • Liquid handler compatible
A 19 µm polymer micro particulate with weak cationic exchange chemistry providing large contac zone for sorbent analyte interaction resulting in a fast and uniform sample prep.