Order Entry
Belgium
ContactUsLinkComponent
Tetra-ubiquitin (K48-linked) (from E. coli)
Tetra-ubiquitin (K48-linked) (from E. coli)
Catalog # ENZOENZPRT2660025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Tetra-ubiquitin (K48-linked) (from E. coli)
Catalog # ENZOENZPRT2660025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Conjugation:
    Unconjugated
  • Source:
    E. coli
  • Seq:
    02
  • Storage conditions:
    –80 °C
  • Protein synonyms:
    Ub4
  • Protein/peptide name:
    Tetra-ubiquitin (K48-linked)
  • Molecular weight:
    ∼34 kDa
  • Formulation:
    Liquid, containing 20 mM Tris-HCl, pH 7,5, 0,15 M NaCl, 1 mM EDTA.
  • Shipping temperature:
    Dry Ice

Specifications

About this item

Produced in E. coli.