Order Entry
Belgium
ContactUsLinkComponent
Screw Closures for Wide Neck Bottle Series 303, PP
Screw Closures for Wide Neck Bottle Series 303, PP
Catalog # KAUT2000794398
Supplier:  Kautex
Screw Closures for Wide Neck Bottle Series 303, PP
Catalog # KAUT2000794398
Supplier:  Kautex

Specifications

  • Thread:
    25 mm
  • Colour:
    Natural
  • Material:
    PP
  • Seq:
    01
  • Pk:
    20
  • For bottles:
    100 ml

Specifications

About this item

Leakproof polypropylene screw closures are packaged in polyethylene bags.