Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microsystems™ Schott Gooseneck Light Guide

Microsystems™ Schott Gooseneck Light Guide

Gooseneck light guide, Microscope
Catalog # LEIM31154202
Supplier:   LEICA MICROSYSTEMS
Microsystems™ Schott Gooseneck Light Guide
Catalog # LEIM31154202
Supplier:  LEICA MICROSYSTEMS
Microsystems™ Schott Gooseneck Light Guide

Gooseneck light guide, Microscope

Price
494,68 €
Log in for availability
1 * 1 ST
Product Details & Documents

High quality schott fiber optics for perfect light placement. Microsystems™ Schott Gooseneck Light Guide provides flex and stay lighting.

  • Precise positioning offers complete control of light placement
  • Gooseneck branches away from light for near shadow-free illumination
  • Easily adjustable, but holds its position
  • Length: 600 mm
  • Diameter : 4,5 mm

Delivery information: Includes with two arms.

Specifications
  • Description:
    Gooseneck light guide
  • For:
    Microscope
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.