High-Power Electronic Crimpers and Decappers
Ergonomic electronic high power crimping tools can effortlessly crimp/decap even the strongest all-steel caps with just the push of a button. The high power unit is designed as a single tool with interchangeable jawsets for the various functions and sizes needed in your laboratory.
- Fixed power supply with cord (no battery)
- Uses multiple jaw sets to achieve various functions and accommodate different size needs
- The controls to increase or decrease crimp settlings are conveniently located on top of the unit, preventing accidental adjustment of the settings
- OLED screen for simple operation displays jaw set size, crimp setting and cycle result
- Performance tracking through an internal cycle log
- Strong enough for all-steel caps
Consistently crimp caps to a reproducible seal setting with less physical strain. No need more multiple battery units - one high power unit covers all sizes and functions needed with the jawset attachments.
Recommended applications: High volume or strenuous crimping/decapping applications including all-steel caps and for requirements of multiple cap sizes and functions.
Jawsets available in 8, 11, 13, and 20 mm crimping and decapping.
Frequently Bought Together