Order Entry
Belgium
ContactUsLinkComponent
SPE Plate, 96-well, NBE PWCX
SPE Plate, 96-well, NBE PWCX
Catalog # TCAT675-0051NBE
Supplier:  Tecan
SPE Plate, 96-well, NBE PWCX
Catalog # TCAT675-0051NBE
Supplier:  Tecan

Specifications

  • Description:
    Plate NBE PWCX, 1 ml, 19 µm
  • Phase:
    PWCX
  • Filling:
    5 mg
  • Pk:
    1

Specifications

About this item

Polymeric, weak cation exchange chemistry for strong bases.

  • Ion-exchange
  • Liquid handler compatible

A 19 µm polymer micro particulate with weak cationic exchange chemistry providing large contac zone for sorbent analyte interaction resulting in a fast and uniform sample prep.