Crimpers and Decappers
Supplier: Trajan Scientific and Medical
Certificates
About this item
Crimping tools have advanced the standards of manual crimpers and decappers by addressing all of the deficiencies associated with traditional blocky, metal tools.
- Reliabily seal crimps caps manually with less physical strain
- constructed of tough, light weight plastic that is incredibly durable but weighs much less than metal crimping tools
- Curved handles create a natural grip, greatly improving comfort over metal grips
- Unique bottom pull technology allows for steadier operation in comparison to top-push handle designs
Application: Low volume crimping/decapping applications for aluminium crimp caps.
The adjustment knob is easily viewed and clearly displays + and − symbols along with directional arrows, simplifying the process of adjusting the crimp setting - no tools needed.
No more extra squeeze required. When the adjustment knob touches the body of the crimper, the desired crimp settling has been reached - significantly reducing arm strain and fatigue associated with traditional manual crimping.
Frequently Bought Together