ContactUsLinkComponent
Whatman™ Reaction Folders, Sealable Hybridisation Bags
Whatman™ Reaction Folders, Sealable Hybridisation Bags
  WHAT10483064
undefined
Whatman™ Reaction Folders, Sealable Hybridisation Bags
  WHAT10483064
  • Description:
    Reaction folders
  • Pk:
    50
  • W×L:
    254×203 mm
Whatman reaction folders are sealable bags for hybridisation and incubation reactions.
  • Folders can be sealed with a standard heat-sealer
The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridisation bags.