Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cerex® PSAX SPE Column

Column Cerex PSAX; 10 mg, PSAX, 80 Å, 17 µm, 1 ml, 10 mg
  TCAT615-0101R
 :   Tecan
Cerex® PSAX SPE Column
  TCAT615-0101R
 :  Tecan
Cerex® PSAX SPE Column

Column Cerex PSAX; 10 mg, PSAX, 80 Å, 17 µm, 1 ml, 10 mg

 
 
   
1 * 96 ST
 

Mixed mode reverse-phase with an anion exchanger for acidic compounds.

  • Ion-exchange
  • Liquid handler compatible

A 17 µm polymer micro particulate with strong anionic exchange chemistry providing large contat zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for acidic target compounds.

 
  • Particle Size:
    17 µm
  • Pore size:
    80 Å
  • Capacity:
    1 ml
  • Description:
    Column Cerex PSAX; 10 mg
  • Bed weight:
    10 mg
  • Phase:
    PSAX