Cerex® HPSCX SPE Columns
Supplier: Tecan
Certificates
About this item
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
- Ion-exchange
- Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.
Frequently Bought Together