For a variety of agarose electrophoresis applications, the ultimate in simplicity and convenience in casting agarose gels.
- Eliminates gel leaking issues, no taping required
- Cast two gel sizes: two 5×8,3 cm and one 10,5×8,3 cm gels
- Designed for easy well identification
- Two combs provided for small and large wells
- Supplied with optional power supply
The casting trays are provided with background black strips for high well visibility. The combs provide either 13 and six wells (0,6×0,1 cm well) or 17 and eight wells (0,4×0,1 cm well) for the large and mini gels, respectively. Interlocked ventilated fog free lid allows full view of gel during electrophoresis. Replacement parts are also available including additional combs (MT-C01, MT-C02), gel trays (MT-T01, MT-T02), casting tray (MT-T03) or a complete gel casting set (one casting tray, two combs, one large tray and two small trays) (MT-T04).
Less agarose consumption leads to less running buffer consumption. Multichannel pipette-compatible. Single molded tank. Safety and ventilation lid enables to cast three pieces of agarose gel in one time. No direct heat impact from power supply unit (high temperature capability, 130 °C), easy sample loading. Great indications for gel making and running. No tapes, clamps or springs needed for gel casting.
Light-weight and simple to use, the gel casting tray is designed to eliminate the leak problems common with other gel casting trays.
- Catalog No:
- GENOBT105-A
- GENOBT105-C
- GENOBT105-F
- Gel dimensions:
- 105×83 mm, 50×83 mm
- 105×83 mm, 50×83 mm
- 50×83 mm