Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stage without Temperature Control

Stage without Temperature Control

A1 fixed stage plate (248×204 mm) for DMi8
Catalog # LEIM11522078
Supplier:   LEICA MICROSYSTEMS
Stage without Temperature Control
Catalog # LEIM11522078
Supplier:  LEICA MICROSYSTEMS
Stage without Temperature Control

A1 fixed stage plate (248×204 mm) for DMi8

Price
832,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The fixed stage plate guarantee long-term stability regardless of environmental conditions.

  • High-quality aluminum
  • Ceramic-coated
  • Extremely scratchproof
  • Precisely plane-parallel
  • Three point mounting

Ordering information: The fixed stage plate is supplied with a round 88 mm insert with a 10 mm opening.

Specifications
  • Description:
    A1 fixed stage plate (248×204 mm) for DMi8
  • Cat. No.:
    LEIM11522078
  • Type:
    Fixed stage plate
  • Accessories included:
    88 mm insert ring with an opening of 10 mm
  • Weight:
    1,45 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.