ContactUsLinkComponent
Sentinel XE® Complete PAPR System with Hood
Sentinel XE® Complete PAPR System with Hood
Catalog # ILCDS-3500EU
Supplier:  ILC DOVER
undefined
Sentinel XE® Complete PAPR System with Hood
Catalog # ILCDS-3500EU
Supplier:  ILC DOVER
Specifications
  • Description:
    Sentinel XE® complete PAPR system with hood, CE marked
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Sentinel XE® PAPR (powered air-purifying respirator) with unique clear hood is specifically designed to delivery TH3 level protection to pharmaceutical and biopharmaceutical workers.
  • Improved safety with a 320° field of view panoramic hood
  • Better comfort; no ratcheted head harness
  • Greater airflow capacity delivers 220 LPM airflow within the hood and suit
  • Lightweight blower provides better ergonomics
  • Longer battery life; 12+ hours of service increases productivity
The Sentinel XE® complete PAPR system with hood is available with CE or UKCA marking.
Certifications: Approved to EN 12941/A2:208 TH3 P R SL. TH3 is the highest level of production and performance available under this certification. RoHS compliant per 2011/65/EU.