ContactUsLinkComponent
Near Vertical LED Illumination System, LED3000
Near Vertical LED Illumination System, LED3000
Catalog # LEIM10450656
Supplier:  LEICA MICROSYSTEMS
Near Vertical LED Illumination System, LED3000
Catalog # LEIM10450656
Supplier:  LEICA MICROSYSTEMS
Specifications
  • Description:
    LED3000 NVI vertical incident microscope illuminator
  • Pk:
    1
Specifications
About this item
The Leica LED3000 NVI is ideal for viewing recesses and holes, since the light falls almost vertically on the sample. In contrast to coaxial illumination it is also ideally suited for non-reflective and uneven sample surfaces.
  • Ideal for viewing recesses and cavities
  • Evenly distributed light through 2-point illumination
  • Minimises shadows caused by tools
  • Compact design - great accessibility to the sample
NVI illumination shines almost straight down onto the sample plane. This allows cavities and recesses to be very well illuminated.