ContactUsLinkComponent
Fixed Eyepiece with Pointer for DM500 and DM750
Fixed Eyepiece with Pointer for DM500 and DM750
Catalog # LEIM13613531
Supplier:  LEICA MICROSYSTEMS
Fixed Eyepiece with Pointer for DM500 and DM750
Catalog # LEIM13613531
Supplier:  LEICA MICROSYSTEMS
Specifications
  • Magnification:
    10×/20
  • Description:
    Fixed eyepiece with pointer for DM500 and DM750 (does not accept a reticle)
  • Pk:
    1