ContactUsLinkComponent
ISO Humidity Calibration Certificate
ISO Humidity Calibration Certificate
Catalog # TEST0520.0166
Supplier:  TESTO
ISO Humidity Calibration Certificate
Catalog # TEST0520.0166
Supplier:  TESTO
Specifications
  • Description:
    ISO humidity calibration certificate - with 3 measuring points
  • Pk:
    1
Specifications
About this item
This ISO calibration certificate certifies that your measuring instrument or probe is calibrated. Calibration is carried out according to the ISO standard.
  • Three calibration points: 11,3/50/75,3% RH at +25 °C
  • According to the ISO standard
  • Product/housing material: Paper
  • Product colour: White