Order Entry
Belgium
ContactUsLinkComponent
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Supplier:  OMEGA BIO-TEK
Certificates
MicroElute® Cycle-Pure & Gel Extraction Kit, Omega Bio-tek
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    MicroElute® Cycle-Pure & Gel Extraction Kit

Specifications

Related Information

Frequently Bought Together