Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microscope Slides with Cut Edges

Microscope Slides with Cut Edges

Microscope slides, pack of 1000 slides, White, Glass, 1 mm, 76×50 mm
Catalog # MARI1100290091
Supplier:   MARIENFELD
Microscope Slides with Cut Edges
Catalog # MARI1100290091
Supplier:  MARIENFELD
Microscope Slides with Cut Edges

Microscope slides, pack of 1000 slides, White, Glass, 1 mm, 76×50 mm

Price
Inquire For Price
Log in for availability
1 * 1.000 ST
Product Details & Documents
Specifications
  • Colour:
    White
  • Description:
    Microscope slides, pack of 1000 slides
  • L×W:
    76×50 mm
  • Material:
    Glass
  • Thickness:
    1 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.