Order Entry
Belgium
Orders LinkContactUsLinkComponent
Objective, Achromat, M60 Thread

Objective, Achromat, M60 Thread

Achromat objective, M60 thread, 188,5 mm, Ext.Ø: 58 mm
Catalog # LEIM10450192
Supplier:   LEICA MICROSYSTEMS
Objective, Achromat, M60 Thread
Catalog # LEIM10450192
Supplier:  LEICA MICROSYSTEMS
Objective, Achromat, M60 Thread

Achromat objective, M60 thread, 188,5 mm, Ext.Ø: 58 mm

Price
571,12 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Ø:
    Ext.Ø: 58 mm
  • Magnification:
    0,50×
  • Description:
    Achromat objective, M60 thread
  • Working distance:
    188,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.