ContactUsLinkComponent
Lightning-Link® Rapid DyLight 450 Antibody Labeling Kits
  NOVU321-0010
 :  
Lightning-Link® Rapid DyLight 450 Antibody Labeling Kits
  NOVU321-0010
 :  
  • Pk:
    1 KIT
  • Description:
    Lightning-link rapid dylight 450 antibody labeling kit, unit size: 3×100 µg reaction
Lightning-link rapid is an innovative technology that enables direct labeling of proteins, peptides or other biomolecules for use in R&D applications, drug discovery and the development of diagnostic kits.
  • Flexible quick and easy to use
  • Fully scalable (10 µg to 1 g or more)
  • 100% recovery - no antibody/protein loss
  • store at −20 °C
DyLight 405 provides blue fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 402 nm, high fluorescence at 428 nm (extinction coefficient 3,0×104 cm-1M-1) and high quantum yield.