Order Entry
Belgium
ContactUsLinkComponent
System Calibration Standard (7 components)
System Calibration Standard (7 components)
Catalog # ACSDF237594
Supplier:  CPACHEM
CAS Number:  
System Calibration Standard (7 components)
Catalog # ACSDF237594
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    5 mL
  • Description:
    System calibration standard - 7 components in n-heptane

Specifications

About this item

System calibration standard with 7 components.

  • Solvent: n-Heptane
  • Volume: ampoule 5 ml

The components consists of cyclohexane 1% (w/v), 1-phenyldodecane 0,1% (w/v), o-Xylene (1,2-dimethylbenzene) 0.5% (w/v), hexamethylbenzene 0,1% (w/v), naphthalene 0,1% (w/v), dibenzothiophene 0,05% (w/v) 9-methylanthracene 0,05% (w/v).

Certifications: EN 12916