Orders LinkContactUsLinkComponent
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code
CR950 reader, with stand and 1,8 m USB cable, W×D×H: 50,8×69,85×139,7 mm
Catalog # BRDY176522
Supplier:   Brady
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code
Catalog # BRDY176522
Supplier:  Brady
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code
CR950 reader, with stand and 1,8 m USB cable, W×D×H: 50,8×69,85×139,7 mm
Price
192,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Improve data collection operations with this swift and accurate barcode reader. The CR950 thrives in fast-paced environments, offering increased efficiency with its intuitive targeting device. This kit features a stand.
  • Swift and accurate barcode reader
  • Kit includes a stand
  • Manual or automatic triggering
  • Provides easier scanning with a wide area image sensor
  • Features two programmable indicators: LED and tone
  • 5-year limited warranty
The CR950 thrives in fast-paced environments, offering increased efficiency with its intuitive targeting device. Get high-speed omnidirectional reading of 1D and 2D barcodes and glare reduction technology for reading barcodes on shiny surfaces.
Specifications
  • Description:
    CR950 reader, with stand and 1,8 m USB cable, W×D×H: 50,8×69,85×139,7 mm