Orders LinkContactUsLinkComponent
TBX agar
TBX agar
TBX agar, 90 mm ready-to-use plates, -, 20
Catalog # LIOF10522
Supplier:   LIOFILCHEM
TBX agar
Catalog # LIOF10522
Supplier:  LIOFILCHEM
CAS Number:  
TBX agar
TBX agar, 90 mm ready-to-use plates, -, 20
CAS Number:  
Price
33,30 €
Log in for availability
1 * 20 ST
Product Details & Documents
X-glucuronide, contained in the TBX agar formulation, is the chromogenic agent that allows the determination of the β-Dglucuronidase activity, which is a highly specific enzyme for E. coli. Gram-positive bacteria are inhibited by bile salts.
Specifications
Specifications
  • Pk:
    20
  • Description:
    TBX agar, 90 mm ready-to-use plates
  • Filling:
Specifications