ContactUsLinkComponent
ClearKlens Solve VH21 Detergent
ClearKlens Solve VH21 Detergent
Catalog # DIVY101103248
Supplier:  Diversey
undefined
ClearKlens Solve VH21 Detergent
Catalog # DIVY101103248
Supplier:  Diversey
Specifications
  • Description:
    ClearKlens solve VH21 detergent
  • Sterility:
    Non sterile
  • pH:
    <11
  • Pk:
    1 Bottle
Specifications
About this item
ClearKlens solve is an alkaline foam detergent for use in the pharmaceutical and cosmetic industries.
  • Balanced blend of anionic and non-ionic surfactants in alkaline solution
  • Removes oils, fats and grease of vegetable and animal origin and other product residues found in the pharmaceutical and cosmetic industries
  • ClearKlens solve is for manual cleaning of process equipment, floors and walls and foam applications
  • Highly effective alkaline detergent for difficult cleaning tasks
  • Can be used on aluminum with care as directed
Use ClearKlens Solve for daily routine cleaning 1 to 2% v/v up to 60 °C, contact time 10 to 15 mins with brushing, followed by rinsing in clean water. Foam applications 3 to 5% v/v. For heavy duty residues requiring repeat treatments up to 10% v/v concentration may be required. ClearKlens Solve containing solutions should be thoroughly rinsed after use to remove them from all product contact surfaces.