Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

MALDI AnchorChip™ Targets

MTP AnchorChip™ 384 BC
Catalog # BRUK8280790
Supplier:   BRUKER DALTRONIK
MALDI AnchorChip™ Targets
Catalog # BRUK8280790
Supplier:  BRUKER DALTRONIK
MALDI AnchorChip™ Targets

MTP AnchorChip™ 384 BC

Price
3 730,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Patented AnchorChip™ targets are polished steel targets with hydrophilic sample and calibrant spots surrounded by a hydrophobic barrier for MALDI sample preparation.

  • Spot Ø: 800 µm
  • Sample spots: 384
  • Calibrant spots: 96
  • Autoteaching

Sample positions on contain 'anchors', hydrophilic patches surrounded by a hydrophobic ring. The 'anchor' localises droplets at the sample position and the hydrophobic ring prevents sample spreading and concentrates the sample into a spot between 0,4 and 2 mm in diameter. The localisation effect ensures that every single laser shot fired throughout an automatic run will hit a sample spot. This significantly increases the efficiency of the MALDI acquisition process.

Compatible with autoflex, ultraflex , rapifleX, timsTOF fleX, scimaX and solariX series instruments.

Specifications
  • Description:
    MTP AnchorChip™ 384 BC
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • MTP Target frame III
    MTP Target frame III
    BRUK8074115
    Log in for availability
    1 * 1 ST - 3 410,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.